Orders LinkContactUsLinkComponent
Slab Easels
Slab Easels
Slab Easels, Slab Easel
Catalog # 470018-778
Supplier:   JULE-ART INC.
Número de proveedor #:  36 A
Slab Easels
Catalog # 470018-778
Supplier:  JULE-ART INC.
Slab Easels
Slab Easels
Slab Easels, Slab Easel
Price
$144.97
Log in Para disponibilidad
Paquete de 4
Product Details & Documents
Safely prop up flat specimens such as agate or fossils in a thin matrix. The smaller easel supports slabs up to 1/4" thick, the larger easel easily accommodates thicker slabs and crystal plates.
Slab Easel Size: 3"L x 2 1/2"W x 2 1/4"H. Package of four.
Thick Slab Easel Size: 2 1/2"L x 2 1/4"W x 2 3/4"H. Package of four.
Especificaciones
  • Description:
    Slab Easels, Slab Easel