United Scientific™ Push/Pull Spring Scales
Push/pull spring scale, green, 500 g/5 N, graduation interval: 0.1 N
Catalog # 470017-446
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: PS0500
United Scientific™ Push/Pull Spring Scales
Catalog # 470017-446
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
Product Details & Documents
Versatile spring scale.
- Weighs in Grams and Newtons
- Color coded to differentiate capacity
These Push Pull Spring Scales are a versatile spring scale that can be used to measure forces using push or pull functions, or to weigh a suspended object from the large hook.
Delivery information: Includes a zero adjustment screw, a metal suspension ring, instructions and application sheets.
Caution: This product is for ages six and up.
Especificaciones
- Description:Push/pull spring scale, green
- Weighing Capacity:500 g/5 N, graduation interval: 0.1 N