Utility Knife
Utility knife replacement blades for 6" knife, pack of 5
Catalog # 470017-316
Supplier: GREAT NECK TOOLS, INC.
Supplier #: 11P
Jump to:
Product Details & Documents
The all-purpose knife stores five heavy-duty alloy-steel blades, included, in the handle. The blade locks in place with a safety slide.
Size: 6"L.
/!\ CAUTION: Contains functional sharp edges.
Specifications
- Description:Utility knife replacement blades for 6" knife, pack of 5