Conductivity Indicator
Conductivity Indicator
Catalog # 470016-796
Supplier: SCIENCE INTERACTIVE GROUP
Número de proveedor #: 611-2260
Jump to:
Product Details & Documents
Light Bulb Glows When Water Conducts Electricity.
- Attaches to a standard ring stand
- Comes with illustrated instructions
Presenting no shock hazard because the cord at the side can’t come in contact with the water, this indicator features a plastic molded body and recessed electrical connections and electrodes. It operates on 110 VAC. A light bulb is required.
Especificaciones
- Description:Conductivity Indicator