Friction Rods
Solid Glass, Rounded
Catalog # 470016-220
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: FRSG12
Jump to:
Product Details & Documents
For your Electrostatic Needs!
- Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
Specifications
- Description:Solid Glass, Rounded