Dissection Aprons
Frog Anatomy Aprons
Catalog # 470014-866
Supplier: RANACO CORP SE
Número de proveedor #: 420.6100.1
Jump to:
Product Details & Documents
Protects and educates.
- Protective and disposable
- Diagrams on aprons help students dissect
Help your students protect their clothing from chemicals and stains and provide them with a dissection guide at the same time. Each 3 mil, polyethylene apron features a black and white illustration of a dissected and labeled fetal pig or frog printed on it. Each pattern comes in a perforated roll of 100 aprons.
Especificaciones
- Description:Frog Anatomy Aprons