Orders LinkContactUsLinkComponent
Boreal2 Microscopes, EM Series
Boreal2 Microscopes, EM Series
Coarse and fine focus intermediate microscope, LED cordless
Catalog # 470014-510
Supplier #:  SK54
Boreal2 Microscopes, EM Series
Catalog # 470014-510
Boreal2 Microscopes, EM Series
Boreal2 Microscopes, EM Series
Coarse and fine focus intermediate microscope, LED cordless
Price
$5,436.68
Log in for availability
Each (1UN)
Product Details & Documents
One of our easiest-to-use models, this is the perfect introductory microscope for students.
  • Full-Size Optics
  • Ergonomic Body Style
  • Features Designed to Prevent Tampering
  • Objectives: 4x, 10x, and 40x
  • Boreal Lifetime Warranty
It boasts a gliding stage with stage clips for one-hand use, focus dots that make focusing quick and easy, and color-coded and distinctly labeled objectives.
The microscope also features an all-metal chassis, illumination without heat through a bright, white LED, and up to 400x magnification.
Ordering information: This product includes a manufacturer's warranty.
Specifications
  • Description:
    Coarse and fine focus intermediate microscope
  • Illumination:
    LED cordless