Boreal2 Microscopes, EM Series
Coarse and fine focus intermediate microscope, LED cordless
Catalog # 470014-510
Supplier: MOTIC HONG KONG LIMITED
Supplier #: SK54
Jump to:
Product Details & Documents
One of our easiest-to-use models, this is the perfect introductory microscope for students.
- Full-Size Optics
- Ergonomic Body Style
- Features Designed to Prevent Tampering
- Objectives: 4x, 10x, and 40x
- Boreal Lifetime Warranty
It boasts a gliding stage with stage clips for one-hand use, focus dots that make focusing quick and easy, and color-coded and distinctly labeled objectives.
The microscope also features an all-metal chassis, illumination without heat through a bright, white LED, and up to 400x magnification.
Ordering information: This product includes a manufacturer's warranty.
Specifications
- Description:Coarse and fine focus intermediate microscope
- Illumination:LED cordless