Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Stackable Banana Plug Connecting Wires

Red Stackable Banana Plug Connecting Wires, 12"
  470014-304
Stackable Banana Plug Connecting Wires
  470014-304
Stackable Banana Plug Connecting Wires
Stackable Banana Plug Connecting Wires

Red Stackable Banana Plug Connecting Wires, 12"

 
$51.16
   
Each (1UN)
 

Flexible, Insulated Wire.

  • Insulated 4 mm banana plug at both ends
  • Plated spring contacts
  • Stack to accept other banana plugs
  • With side connectors
 
  • Description:
    Red Stackable Banana Plug Connecting Wires
  • Length:
    12"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.