Equilateral Acrylic Prisms
Prism, Equilateral/Acrylic, 75 mm x 25 mm
Catalog # 470013-002
Supplier: UNITED SCIENTIFIC SUPPLIES
: PAE075
Jump to:
Product Details & Documents
Demonstrate refraction and internal reflection.
- Highly polished sides
- High optical clarity
Selection of equilaterial and right angle acrylic prisms.
Especificaciones
- Description:Prism, Equilateral/Acrylic, 75 mm x 25 mm