Tandem Pulleys
Tandem Pulleys, Triple
Catalog # 470012-862
Supplier: PRECISION MOULDED POLYMERS LIMITED
: 1350102060003
Jump to:
Product Details & Documents
High Quality and Economical
- Free-running
- Easy mounting
- Economical
These are free-running tandem pulleys with rigid plastic sheaves and nickel-plated steel blocks with a hook at each end for easy mounting. Double 1 1/2" and 2" sheaves.
Specifications
- Description:Tandem Pulleys, Triple