Orders LinkContactUsLinkComponent
Tandem Pulleys
Tandem Pulleys
Tandem Pulleys, Double
Catalog # 470012-860
Número de proveedor #:  1350102050003
Tandem Pulleys
Catalog # 470012-860
Tandem Pulleys
Tandem Pulleys
Tandem Pulleys, Double
Liquidación
$1.95
Oferta
Válido hasta 31/12/2026
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
High Quality and Economical
  • Free-running
  • Easy mounting
  • Economical
These are free-running tandem pulleys with rigid plastic sheaves and nickel-plated steel blocks with a hook at each end for easy mounting. Double 1 1/2" and 2" sheaves.
Especificaciones
  • Description:
    Tandem Pulleys, Double