Assorted Wires
Enameled copper wire, 4 oz., 200 ft., 24
Catalog # 470012-728
Supplier: ARCOR ELECTRONICS CO INC SE
Supplier #: A24-SCIEN
Jump to:
Product Details & Documents
Specifications
- Description:Enameled copper wire, 4 oz.
- Length:200 ft.
- Gauge:24