Orders LinkContactUsLinkComponent
Correlating Sedimentary Strata Kit
Correlating Sedimentary Strata Kit
Correlating Sedimentary Strata Kit, Single Set
Catalog # 470012-492
Número de proveedor #:  443SEL
Correlating Sedimentary Strata Kit
Catalog # 470012-492
Correlating Sedimentary Strata Kit
Correlating Sedimentary Strata Kit
Correlating Sedimentary Strata Kit, Single Set
Price
$734.49
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
View Sequence of Rock Layers In Different Locations
Illustrate the law of superposition in determining the relative ages of rock layers. In this activity, four simulated drill cores represent a series of rock layers from different locations. Students use the evidence from each drill core to create a stratigraphic column for each locality. Then, using the fossils within each layer, students learn that similar sequences of rock layers, including the fossils within them, are found in different geographical areas despite their distance from each other. Students are also challenged to construct a timeline showing the relative time spans of each fossil found. Includes a teacher’s guide and student worksheets.
Especificaciones
  • Description:
    Correlating Sedimentary Strata Kit, Single Set