Orders LinkContactUsLinkComponent
Viable Pea Seeds
Viable Pea Seeds
Wrinkled Seeds, 16 oz.
Catalog # 470007-182
Supplier #:  820LB
Viable Pea Seeds
Catalog # 470007-182
Viable Pea Seeds
Viable Pea Seeds
Wrinkled Seeds, 16 oz.
Price
$138.60
Log in for availability
Paquete de 1
Product Details & Documents
Learn genetics like Mendel.
  • Seeds of multiple Phenotypes
  • Variable sizes for each seed type
Use these peas in any lab activity, not just genetics. Peas are an excellent option for germination, growth, and genetic experiments.
Accessories information: Where a Count of Seeds is indicted, the Quantity is an estimate based on Weight and may vary slightly.
Specifications
  • Description:
    Wrinkled Seeds
  • Size:
    16 oz.