Balls for Physics Experiments
Drilled Hardwood Ball, 25 mm, 1"
Catalog # 470007-142
Supplier: EISCO SCIENTIFIC LLC
Supplier #: HWDBL25
Jump to:
Product Details & Documents
Ideal for Collision and Pendulum Demonstrations
Drilled balls for pendulum bobs or impact experiments.
Specifications
- Description:Drilled Hardwood Ball, 25 mm
- Diameter:1"