Orders LinkContactUsLinkComponent
Pendulum Clamp
Pendulum Clamp
Catalog # 470007-140
Pendulum Clamp
Catalog # 470007-140
Supplier:  
Pendulum Clamp
Pendulum Clamp
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BALL BRASS DRILLED .75IN / 19MM D Catalog #470137-640
Product 470137-640 is similar to 470007-140 but may have a different manufacturer and/or packaging.
Product Details & Documents
This sturdy clamp is invaluable for experiments involving energy, motion, acceleration, and more.
  • Clamp Hangs Three Pendulums at Once
  • Easy-To-Adjust Pendulum Lengths
  • Length: 10⅝"
  • Topics Include Energy, Motion, Acceleration, and More
It hangs three pendulums at once, and you can adjust any pendulum without disturbing the others. The support arm screws into the included rod clamp and has thumbscrews for suspension cords.
10⅝" in length; it can be used with a 1/2" or 3/4" dia. vertical support rod.