Glass Cylindrical Mirrors
Convex/cylindrical mirror
Catalog # 470007-094
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MRCV02
Jump to:
Product Details & Documents
Demonstrate converging and diverging mirrors.
- Self standing
- Glass with smooth edges
Both are perfect for use in all your light experiments. Both measure 25 x 50 mm and have a focal length of 75 mm.
Especificaciones
- Description:Convex/cylindrical mirror