Orders LinkContactUsLinkComponent
Glass Cylindrical Mirrors
Glass Cylindrical Mirrors
Convex/cylindrical mirror
Catalog # 470007-094
Número de proveedor #:  MRCV02
Glass Cylindrical Mirrors
Catalog # 470007-094
Glass Cylindrical Mirrors
Glass Cylindrical Mirrors
Convex/cylindrical mirror
Price
$103.94
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Demonstrate converging and diverging mirrors.
  • Self standing
  • Glass with smooth edges
Both are perfect for use in all your light experiments. Both measure 25 x 50 mm and have a focal length of 75 mm.
Especificaciones
  • Description:
    Convex/cylindrical mirror