Glass Cylindrical Mirrors
Concave/cylindrical mirror
470007-092
: MRCC01
Demonstrate converging and diverging mirrors.
- Self standing
- Glass with smooth edges
Both are perfect for use in all your light experiments. Both measure 25 x 50 mm and have a focal length of 75 mm.
- Description:Concave/cylindrical mirror