Orders LinkContactUsLinkComponent
Glass Cylindrical Mirrors
Concave/cylindrical mirror
  470007-092
 :  MRCC01
Glass Cylindrical Mirrors
  470007-092
Glass Cylindrical Mirrors
Glass Cylindrical Mirrors
Concave/cylindrical mirror
$103.94
Each (1UN)
Demonstrate converging and diverging mirrors.
  • Self standing
  • Glass with smooth edges
Both are perfect for use in all your light experiments. Both measure 25 x 50 mm and have a focal length of 75 mm.
  • Description:
    Concave/cylindrical mirror