Orders LinkContactUsLinkComponent
Support Stand with Rod
Support Stand with Rod
15×23 cm (Base), 610 mm
Catalog # 470005-862
Supplier #:  SSB6X9-H
Support Stand with Rod
Catalog # 470005-862
Support Stand with Rod
Support Stand with Rod
15×23 cm (Base), 610 mm
Price
$562.33
Log in for availability
Each (1UN)
Product Details & Documents
Weighted With Additional Steel Plate
  • Zinc plated rod
These stamped steel bases feature an additional steel plate on the back, which makes them 50% heavier than regular bases. The steel plate also facilitates a sturdier threading of the rod into the base.
Specifications
  • Rod Length:
    610 mm
  • Dimensions:
    15×23 cm (Base)