Orders LinkContactUsLinkComponent
Support Stand with Rod
13×20 cm (Base), 510 mm
Catalog # 470005-860
 :  SSB5X8-H
Support Stand with Rod
Catalog # 470005-860
Support Stand with Rod
Support Stand with Rod
13×20 cm (Base), 510 mm
Price
$420.00
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Weighted With Additional Steel Plate
  • Zinc plated rod
These stamped steel bases feature an additional steel plate on the back, which makes them 50% heavier than regular bases. The steel plate also facilitates a sturdier threading of the rod into the base.
Especificaciones
  • Rod Length:
    510 mm
  • Dimensions:
    13×20 cm (Base)