Order Entry
Mexico
Orders LinkContactUsLinkComponent
Unmounted Mirrors

Unmounted Mirrors

Convex mirror, 76 mm, 75 mm
Catalog # 470005-648
Número de proveedor #:  MCV703
Unmounted Mirrors
Catalog # 470005-648
Unmounted Mirrors
Unmounted Mirrors

Convex mirror, 76 mm, 75 mm

Price
$92.21
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Converging and diverging mirrors for optical experiments.

  • Optically true glass
  • Scratch-resistant copper back silvering

An excellent selection of spherical, silvered glass mirrors for demonstrations and experiments in optics.

Especificaciones
  • Description:
    Convex mirror
  • Focal Length:
    76 mm
  • Diameter:
    75 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.