Friction Rods
Solid Acrylic, Flat Ended
Catalog # 470005-504
Supplier: LAIRD PLASTICS INC CP
Número de proveedor #: PER QUOTE
Jump to:
Product Details & Documents
For your Electrostatic Needs!
- Excellent quality
Friction rods are used for producing static charges when rubbed against materials such as wool, flannel, silk, and fur.
Especificaciones
- Description:Solid Acrylic, Flat Ended