C Clamps
8 cm (3") Jaw Opening
Catalog # 470005-368
Supplier: UNITED SCIENTIFIC SUPPLIES
: CLMPC3
Jump to:
Product Details & Documents
Securely hold two flat object together.
- Cast iron frame
- Screw thread mechanism
These cast iron C clamps help to securely holes surfaces together. The maximum opening of the jaw needs to be taken into account when selecting the correct size. The horizontal torque bar allows you to apply a lot of force to tighten or release.
Especificaciones
- Description:8 cm (3") Jaw Opening