Orders LinkContactUsLinkComponent
C Clamps
8 cm (3") Jaw Opening
Catalog # 470005-368
 :  CLMPC3
C Clamps
Catalog # 470005-368
C Clamps
C Clamps
8 cm (3") Jaw Opening
Price
$165.77
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Securely hold two flat object together.
  • Cast iron frame
  • Screw thread mechanism
These cast iron C clamps help to securely holes surfaces together. The maximum opening of the jaw needs to be taken into account when selecting the correct size. The horizontal torque bar allows you to apply a lot of force to tighten or release.
Especificaciones
  • Description:
    8 cm (3") Jaw Opening