Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ohaus® Spring Scales

Ohaus® Spring Scales

8002-MN - 20 g (0.2 N) graduation intervals, 500 g (5 N)
Catalog # 470005-238
Supplier:   OHAUS CORP.
Número de proveedor #:  80000015
Ohaus® Spring Scales
Catalog # 470005-238
Supplier:  OHAUS CORP.
Ohaus® Spring Scales
Ohaus® Spring Scales

8002-MN - 20 g (0.2 N) graduation intervals, 500 g (5 N)

Price
$938.10
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

These spring scales are designed to stand up to heavy student use.

  • Scales withstand sudden load release
  • Individually calibrated
  • Color-coded
  • High-contrast markings

Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.

Scales are color coded, and high-contrast markings make readings easy.

Especificaciones
  • Description:
    8002-MN - 20 g (0.2 N) graduation intervals
  • Weighing Capacity:
    500 g (5 N)