Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ohaus® Spring Scales

Ohaus® Spring Scales

8001-MN - 10 g (0.1 N) graduation intervals, 250 g (2.5 N)
Catalog # 470005-236
Supplier:   OHAUS CORP.
Supplier #:  80000014
Ohaus® Spring Scales
Catalog # 470005-236
Supplier:  OHAUS CORP.
Ohaus® Spring Scales
Ohaus® Spring Scales

8001-MN - 10 g (0.1 N) graduation intervals, 250 g (2.5 N)

Price
$938.10
Log in for availability
Each (1UN)
Product Details & Documents

These spring scales are designed to stand up to heavy student use.

  • Scales withstand sudden load release
  • Individually calibrated
  • Color-coded
  • High-contrast markings

Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.

Scales are color coded, and high-contrast markings make readings easy.

Specifications
  • Description:
    8001-MN - 10 g (0.1 N) graduation intervals
  • Weighing Capacity:
    250 g (2.5 N)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.