Sliding Microtome
Replacement Blade for Sliding Microtome, Set/3
Catalog # 470005-084
Supplier: UCHIDA YOKO GLOBAL CO., LTD
Número de proveedor #: 916-2304
Jump to:
Product Details & Documents
Hands-on tool for scientific studies
- Built in safety features
- Adjustable section thickness
Cut sections to 40-50 microns safely and easily with this microtome, which features a clear plastic safety cover, fixed blade and rugged plastic construction for hands-on student use.
Simply secure the specimen in the holder, advance using the feed screw, then draw it over the blade. The feed screw moves the specimen only 0.5 mm per complete rotation, and can be advanced as little as 10 microns. For very thin sections, the specimen can be wrapped in an artificial pith.
Also Includes:
Three, two-sided replacement blades
Artificial piths
Especificaciones
- Description:Replacement Blade for Sliding Microtome, Set/3