Bottle Rocket Launcher
Dual Bottle Rocket Launcher
Catalog # 470003-790
Supplier: SCIENCE INTERACTIVE GROUP
Número de proveedor #: 17010
Jump to:
Product Details & Documents
A Safe and Easy Alternative to Engine Rockets
Take your students outside to show them a dramatic demonstration of the laws of motion. With the simple materials in the bottle rocket kit, any standard plastic soda bottle that you provide up to two liters in size, can be launched into the air. It comes with operating instructions and suggested experiments. Stand size: 8"L x 2"W x 1"H, Length of air line: 5'; length of trigger line: 10'.
Note: Base launcher not included.
Especificaciones
- Description:Dual Bottle Rocket Launcher
Accessories
Información del producto
- Catalog #HeightAvailabilityPrice
- Bicycle Pump with Ø 11/4"