Polyethylene Dispensing Bottles
Bottle, Dispensing, 59 mL
Catalog # 470003-782
Supplier: PLASTIC BOTTLES INC.
Número de proveedor #: 301 & 351
Jump to:
Product Details & Documents
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.
Especificaciones
- Description:Bottle, Dispensing, 59 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



