Twistit® Rubber Stoppers
Stopper, Rubber, Twistit, Size 7, Black, 3 Holes
Catalog # 470003-668
Supplier: PLASTICOID CO INC THE SE
Número de proveedor #: J7--M299
Jump to:
Product Details & Documents
An outstanding advance in rubber stoppers, four stoppers in one reduce inventory confusion and cost.
- Twist one of the rubber protrusions off and the solid stopper becomes a one-hole stopper
- Twist the second one off for a two-hole stopper
- Twist the third one off for a three-hole stopper
Made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation.
Especificaciones
- Description:Stopper, Rubber, Twistit, Size 7, Black, 3 Holes