Orders LinkContactUsLinkComponent
Twistit® Rubber Stoppers
Twistit® Rubber Stoppers
Stopper, Rubber, Twistit, Size 7, Black, 3 Holes
Catalog # 470003-668
Número de proveedor #:  J7--M299
Twistit® Rubber Stoppers
Catalog # 470003-668
Twistit® Rubber Stoppers
Stopper, Rubber, Twistit, Size 7, Black, 3 Holes
Price
$1,155.51
Log in Para disponibilidad
Paquete de 40 Case de 10 (40UN)
Product Details & Documents
An outstanding advance in rubber stoppers, four stoppers in one reduce inventory confusion and cost.
  • Twist one of the rubber protrusions off and the solid stopper becomes a one-hole stopper
  • Twist the second one off for a two-hole stopper
  • Twist the third one off for a three-hole stopper
Made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation.
Especificaciones
  • Description:
    Stopper, Rubber, Twistit, Size 7, Black, 3 Holes