Ward's Portable Electronic Balances
These economical balances feature a stackable design with a shipping load cell lock and overload protection for up to 200 pounds.
- Gram and Newton Measurement
- Balances Powered by AC Power Adapter (Included) or Four AA Batteries (not Included)
- One-Year Manufacturer's Warranty
- Stackable Design
- Shipping Load Cell Lock
They feature the same interface capabilities as the Scout® Pro.
The flip top also acts as a protective stack cover in addition to providing complete draft protection. The top cover center opening functions as a sample tray when removed and turned upside down.
Each balance is designed with a flip-top draft shield to access the chamber through an innovative center opening.
2 part plug available as replacement: 470020-496 goes to back of balance and it has cable. 470020-552 is 2 prong plug which goes to the wall socket. Customer will need both these parts for the balance to operate.
: R232 and USB cables and connectors available separately.
: One-year manufacturer's warranty is included with purchase.
- Pan/Platform Size:Ø 120 mm
- Repeatabilty:0.01 g
- Linearity:0.01
- Readability:0.01 g
- Weighing Capacity:150 g



