Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Dissection Placemats

Frog Dissection Placemat, Polyvinyl, 40 cm, 24.7 cm
Catalog # 470002-394
Supplier:   RANACO CORP SE
 :  420.5030.1
Dissection Placemats
Catalog # 470002-394
Supplier:  RANACO CORP SE
Dissection Placemats
Dissection Placemats

Frog Dissection Placemat, Polyvinyl, 40 cm, 24.7 cm

Price
$291.03
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Perfect Instructional Companion for Wet Lab Areas

  • Resistant to spills of any kind

Illustrations and descriptive text are printed right on the durable polyvinyl mat, so students can have specimen classification and information on the internal and external anatomy right on the lab bench with them while they are dissecting. Can be easily cleaned with a sponge or a paper towel, so students don’t have to worry about damaging a text book or other paper copies.

Especificaciones
  • Width:
    24.7 cm
  • Description:
    Frog Dissection Placemat
  • Length:
    40 cm
  • Finish:
    Polyvinyl