Economy Dissection Instruments
Dissecting Scissors, Nickel Plated, 110 mm (4.5"), Sharp/Sharp
Catalog # 470001-828
Supplier: DUNRITE INSTRUMENTS SE
Número de proveedor #: 140525
Jump to:
Product Details & Documents
Priced for even the tightest budget.
- Plated steel
- Available in multiple sizes and styles
- Great for student dissection sets
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Dissecting Scissors
- Length:110 mm (4.5")
- Material:Nickel Plated
- Tip Style:Sharp/Sharp