Orders LinkContactUsLinkComponent
Economy Dissection Instruments
Economy Dissection Instruments
Dissecting Scissors, Nickel Plated, 110 mm (4.5"), Sharp/Sharp
Catalog # 470001-828
Número de proveedor #:  140525
Economy Dissection Instruments
Catalog # 470001-828
Economy Dissection Instruments
Economy Dissection Instruments
Dissecting Scissors, Nickel Plated, 110 mm (4.5"), Sharp/Sharp
Price
$18.66
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Priced for even the tightest budget.
  • Plated steel
  • Available in multiple sizes and styles
  • Great for student dissection sets
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
  • Description:
    Dissecting Scissors
  • Length:
    110 mm (4.5")
  • Material:
    Nickel Plated
  • Tip Style:
    Sharp/Sharp