Order Entry
Mexico
Orders LinkContactUsLinkComponent
Rudiger Human Systems Charts

Rudiger Human Systems Charts

Lymphatic System Chart, Laminated, 100 cm, 70 cm
Catalog # 470023-186
Número de proveedor #:  CH 9
Rudiger Human Systems Charts
Catalog # 470023-186
Rudiger Human Systems Charts
Rudiger Human Systems Charts

Lymphatic System Chart, Laminated, 100 cm, 70 cm

Price
$485.03
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Anatomical Systems Illustrated from Various Perspectives.

  • Printed on heavy stock and laminated for durability

This series of visually arresting charts displays the major human anatomical systems using full-color, beautifully detailed drawings, allowing students to see each system in depth. Individual structures on each chart are fully identified.

The Complete Series with Flip Chart head and Tripod Stand includes the following charts: Skeleton, Muscular System, Nervous System, Vascular System, Spine, Lower Limb, Lymphatic System, Eye, Heart, Upper Limb, Brain, Respiratory System, Digestive Tract, Pregnancy and Birth, and Head and Neck.

Especificaciones
  • Width:
    70 cm
  • Description:
    Lymphatic System Chart
  • Length:
    100 cm
  • Finish:
    Laminated