Pocket Magnifiers
Pocket Magnifier Double Lens
Catalog # 470021-804
Supplier: PINETREE INTRUMENTS SE
Supplier #: 23202-3
Jump to:
Product Details & Documents
These pocket magnifiers are economical enough to outfit an entire class.
- Handy plastic magnifiers
- Ideal for younger students
- Folds conveniently into attached case
- Double/triple lenses can be stacked to increase magnification
Handy plastic magnifiers, with your choice of single, double, or triple lenses, are ideal for younger students. Each folds conveniently into an attached oval case. The 10X and 15X models’ double and triple lenses can be used separately or “stacked” to increase the magnification.
Dimensions: 1 1/2L x 1 1/4W x 1/4"H folded, 2 3/4"L extended.
Specifications
- Magnification:5×, 10×
- Description:Pocket Magnifier Double Lens