Replacement Bulbs
Replacement Bulb, with Bayonet Base, 110 V, 20 W
Catalog # 470021-694
Supplier: WALTER PRODUCTS SE
Supplier #: LBB1
Jump to:
Product Details & Documents
Replacement microscope light bulbs are available for a variety of microscopes. If unsure which bulb type is needed, please contact Ward's Science.
Especificaciones
- Description:Replacement Bulb, with Bayonet Base
- Electrical:110 V, 20 W