Orders LinkContactUsLinkComponent
Polypropylene Beaker Sets
Polypropylene Beaker Sets
50 / 100 / 250 / 500 / 1000 ml, Set of 5, 5 / 10 / 25 / 50 / 100 ml
Catalog # 470020-818
Supplier:   DYNALON
Número de proveedor #:  522135-0001
Polypropylene Beaker Sets
Catalog # 470020-818
Supplier:  DYNALON
Polypropylene Beaker Sets
Polypropylene Beaker Sets
50 / 100 / 250 / 500 / 1000 ml, Set of 5, 5 / 10 / 25 / 50 / 100 ml
Price
$1,022.31
Log in Para disponibilidad
Kit de 5
Product Details & Documents
Unbreakable Polypropylene
  • Autoclavable
  • Chemical-resistant
These beaker sets are autoclavable up to 145°C and chemical resistant. All feature pour spouts and metric graduation scales.
Especificaciones
  • Graduation Interval:
    5 / 10 / 25 / 50 / 100 ml
  • Capacity:
    50 / 100 / 250 / 500 / 1000 ml
  • Description:
    Set of 5