Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Alkaline Batteries

Alkaline battery 1.5 V, C, 1.5 V
  470019-928
 :  ALK-C PANASONIC
Alkaline Batteries
  470019-928
Alkaline Batteries
Alkaline Batteries

Alkaline battery 1.5 V, C, 1.5 V

 
$74.08
   
Each (1UN)
 

Alkaline batteries available in 9 V, C, D, AA types.

 
  • Voltage:
    1.5 V
  • Description:
    Alkaline battery 1.5 V
  • Type:
    C