Disposable Micropipet Tips
200 - 1000 µl, Blue, 1000 Tips/Bag
Catalog # 470019-832
Supplier: FISCHER TECHNICAL CO SE
Número de proveedor #: D592
Jump to:
Product Details & Documents
High quality micropipette tips to meet any lab's needs.
- Certified DNase, RNase and pyrogen free
- Material: PP
- Autoclavable
- Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
Especificaciones
- Volume:200 - 1000 µl
- Tip Color:Blue
- Packed:1000 Tips/Bag