Environmentally Friendly Thermometers
Environmentally Friendly Thermometers, 1 °C, Partial Immersion, −20...+110 °C, 12"
Catalog # 470019-642
Supplier: THERMCO PRODUCTS INC SE
: B110CY3BLK
Jump to:
Product Details & Documents
Glass thermometers with partial or total immersion.
Especificaciones
- Description:Environmentally Friendly Thermometers, 1 °C, Partial Immersion
- Length:12"
- Range:−20...+110 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



