Orders LinkContactUsLinkComponent
Wire Gauzes, Plain
Wire Gauze, Plain, 4" x 4", 10×10 cm (4×4")
  470019-498
 :  WGPL44-PK/12
Wire Gauzes, Plain
  470019-498
 :  
Wire Gauzes, Plain
Wire Gauzes, Plain
Wire Gauze, Plain, 4" x 4", 10×10 cm (4×4")
 :  
$236.66
Paquete de 12
Sturdy wire mesh for supporting glassware over a Bunsen burner.
  • Long lasting, heat- and tear-resistant iron
  • Available in a variety of sizes
Sheet of cross-hatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time. Edges are folded over for a clean perimeter surface.
  • Description:
    Wire Gauze, Plain, 4" x 4"
  • Size:
    10×10 cm (4×4")