Curved Microdissection Forceps
Curved Microdissection Forceps, Stainless steel, 108 mm (4.25"), Curved
Catalog # 470018-962
Supplier: DUNRITE INSTRUMENTS SE
Número de proveedor #: 141010
Jump to:
Product Details & Documents
Micro fine, curved or straight points enhance control when probing the most delicate structures.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Curved Microdissection Forceps
- Length:108 mm (4.25")
- Material:Stainless steel
- Tip Style:Curved