Science Take-Out® Mark-Recapture Population Sampling
Mark-Recapture population sampling single kit, 1
Catalog # 470017-740
Supplier: SCIENCE TAKE-OUT, LLC
Supplier #: 111
Science Take-Out® Mark-Recapture Population Sampling
Catalog # 470017-740
Supplier: SCIENCE TAKE-OUT, LLC
Jump to:
Product Details & Documents
How Many Spiders Are Hiding In The Leaf Litter?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
How many plastic spiders are hiding in simulated “leaf litter”? Use the mark-recapture population sampling technique to estimate the number of organisms in a population. Capture, mark and release a random sample of spider in your "leaf litter". Recapture a second random sample of spiders and use a mathematical equation to estimate the total number of animals in the population. This complete "dry lab" activity contains all required materials.
Documents
Specifications
- Description:Mark-Recapture population sampling single kit
- No. of Lab Groups:1