Orders LinkContactUsLinkComponent
Economical Class 7 Hooked Weights
Hooked Weight, 1000 g
  470017-686
 :  30390786
Economical Class 7 Hooked Weights
  470017-686
Economical Class 7 Hooked Weights
Economical Class 7 Hooked Weights
Hooked Weight, 1000 g
$2,910.21
Each (1UN)
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
  • Weights above 5 g are constructed of 303 Stainless Steel
  • Weights 5 g and below are constructed of 3003-H14 Aluminum
  • Manufactured in the USA
All weights are hooked and feature two-piece construction. Weight sets are supplied in a weight block with compartments for each weight. Individual weights are not supplied in a case.
  • Description:
    Hooked Weight, 1000 g