Economical Class 7 Hooked Weights
Hooked Weight, 1000 g
470017-686
: 30390786
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
- Weights above 5 g are constructed of 303 Stainless Steel
- Weights 5 g and below are constructed of 3003-H14 Aluminum
- Manufactured in the USA
All weights are hooked and feature two-piece construction. Weight sets are supplied in a weight block with compartments for each weight. Individual weights are not supplied in a case.
- Description:Hooked Weight, 1000 g