LaMotte® Soil Texture Kit
With the Soil Texture Kit, students determine the percentage of sand, silt, and clay in soil samples.
- Students Separate Different Sizes of Particles
- For Use in the Field or the Lab
- Instructions and Study Guide Included
- Kit Includes Material for 50 Tests
- Texture-Interpretation Guide Helps Students Decode Results
They can use the kit right in the field to see how sand, silt, and clay affect growth in the root zone.
The procedure allows them to separate the substances from the soil, determine their respective percentages in soil samples, and decode the results with an easy-to-read texture-interpretation guide.
: This kit includes materials for 50 tests, a soil study guide, and instructions.
- Description:Refill Kit for Soil Texture Kit, 50 tests
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.