Orders LinkContactUsLinkComponent
Economical Class 7 Hooked Weights
Hooked Weight, 20 g
  470013-438
 :  30390791
Economical Class 7 Hooked Weights
  470013-438
Economical Class 7 Hooked Weights
Economical Class 7 Hooked Weights
Hooked Weight, 20 g
$18.10
  31/12/2026
Each (1UN)
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
  • Weights above 5 g are constructed of 303 Stainless Steel
  • Weights 5 g and below are constructed of 3003-H14 Aluminum
  • Manufactured in the USA
All weights are hooked and feature two-piece construction. Weight sets are supplied in a weight block with compartments for each weight. Individual weights are not supplied in a case.
  • Description:
    Hooked Weight, 20 g