Order Entry
Mexico
Orders LinkContactUsLinkComponent
Economical Class 7 Hooked Weights

Economical Class 7 Hooked Weights

Hooked Weight, 50 g
Catalog # 470013-390
Supplier:   TROEMNER LLC
Supplier #:  30390790
Economical Class 7 Hooked Weights
Catalog # 470013-390
Supplier:  TROEMNER LLC
Economical Class 7 Hooked Weights
Economical Class 7 Hooked Weights

Hooked Weight, 50 g

Price
$767.53
Log in for availability
Each (1UN)
Product Details & Documents

For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.

  • Weights above 5 g are constructed of 303 Stainless Steel
  • Weights 5 g and below are constructed of 3003-H14 Aluminum
  • Manufactured in the USA

All weights are hooked and feature two-piece construction. Weight sets are supplied in a weight block with compartments for each weight. Individual weights are not supplied in a case.

Specifications
  • Description:
    Hooked Weight, 50 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.