Orders LinkContactUsLinkComponent
Variable Speed Centrifuge
Variable Speed Centrifuge
Centrifuge Adapters for 3.0-7.5mL Short Tubes
Catalog # 470012-612
Supplier #:  C800-03
Variable Speed Centrifuge
Catalog # 470012-612
Variable Speed Centrifuge
Variable Speed Centrifuge
Centrifuge Adapters for 3.0-7.5mL Short Tubes
Price
$1,498.82
Log in for availability
Paquete de 10
Product Details & Documents
A Valuable Addition to Any Laboratory.
  • 30 minute timer with bell tone signal
  • Latching lid with Safety Switch
  • Easily accessible stainless steel rotor
For a clean, complete separation of cellular components yielding a superior serum or plasma specimen, use this powerful variable speed centrifuge. Suction cup rubber feet for stability also reduce vibration. Centrifuge features all UL approved components.
Specifications
  • Description:
    Centrifuge Adapters for 3.0-7.5mL Short Tubes