Order Entry
Mexico
Orders LinkContactUsLinkComponent
LaMotte® pH TRACER

LaMotte® pH TRACER

LaMotte® pH TRACER
Catalog # 470012-552
Número de proveedor #:  1741
LaMotte® pH TRACER
Catalog # 470012-552
LaMotte® pH TRACER
LaMotte® pH TRACER

LaMotte® pH TRACER

Price
$5,652.54
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Store Up to 15 Readings

  • Automatic Temperature Compensation
  • Auto-off to Conserve Battery Life
  • Rugged Flat Electrode

For clear readings everytime, this pH meter features a large, bold display, which shows pH and temperature of a sample in numbers or bar graphs. A built-in memory stores up to 15 readings. Automatic shut-off and low battery indicator are additional features. It includes 4, 7, and 10 pH buffer tablets and four LR-44 batteries. pH range: 0–14, resolution: 0.01 pH, accuracy: ±0.01 pH. Temperature range: 32–149°F (0–65°C), resolution: 0.1°F/C.

pH Range: 0 - 14. Resolution: 0.01 Accuracy: ±0.01 pH.
Temperature Range: 32–149 °F; 0–65 °C. Resolution: 0.1°F/°C.

Especificaciones
  • Description:
    LaMotte® pH TRACER
  • Cat. No.:
    470012-552
  • pH accuracy:
    ±0.01
  • pH resolution:
    0.01
  • pH range:
    0 - 14
  • Temperature range:
    32–149 °F; 0–65 °C
  • Temperature resolution:
    0.1°F/°C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.