BioCam Cell Structure And Function Charts
Cell Wall Chart, Laminated, 61 cm, 45.7 cm
Catalog # 470006-418
Supplier: BIOCAM COMMUNICATIONS INC SE
: WC 52
BioCam Cell Structure And Function Charts
Catalog # 470006-418
Supplier: BIOCAM COMMUNICATIONS INC SE
Jump to:
Product Details & Documents
A mixture of photo-micrographs, detailed artwork and diagrams make these charts excellent for supporting your unit on cells and cell division.
- Highly detailed
Explore cells with these charts.
Especificaciones
- Width:45.7 cm
- Description:Cell Wall Chart
- Length:61 cm
- Finish:Laminated