Biocam Pathogen Charts
Microbial Food Poisoning Chart, Laminated, 61 cm, 45.7 cm
Catalog # 470006-410
Supplier: BIOCAM COMMUNICATIONS INC SE
Número de proveedor #: WC54
Jump to:
Product Details & Documents
These high-resolution microphotography charts will make an excellent addition to the walls of your classroom.
- Provide superior definition of structures without the aid of a microscope
Especificaciones
- Width:45.7 cm
- Description:Microbial Food Poisoning Chart
- Length:61 cm
- Finish:Laminated